Merck
  • Efficient Syntheses of Novel Fluoro-Substituted Pentacenes and Azapentacenes: Molecular and Solid-State Properties.
Efficient Syntheses of Novel Fluoro-Substituted Pentacenes and Azapentacenes: Molecular and Solid-State Properties.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-08-08)
Jonas Schwaben, Niels Münster, Michael Klues, Tobias Breuer, Philipp Hofmann, Klaus Harms, Gregor Witte, Ulrich Koert
ABSTRACT
Non-symmetrical 6,13-disubstituted pentacenes bearing trifluoromethyl and aryl substituents have been synthesized starting from pentacenequinone. Diazapentacenes with a variety of fluorine substituents were prepared either via a Hartwig-Buchwald aryl amination route or by a SNAr strategy. As a result of a non-symmetric substitution pattern containing electron-donating substituents in combination with electron-accepting fluorine substituents, the synthesized compounds feature distinct molecular dipoles. All compounds are analyzed regarding their optoelectronic properties in solution with special focus on the frontier orbital energies as well as their molecular packing in the crystal structures. The analyses of isolated molecules are complemented by thin-film studies to examine their solid-state properties. A precise comparison between these and the molecular properties gave detailed insights into the exciton binding energies of these compounds, which are explained by means of a simple model considering the molecular packing and polarizabilities.
Sigma-Aldrich
RuPhos Pd G2
Sigma-Aldrich
RuPhos Pd G2 Chembeads