Merck
  • Xepac protein and IP3/Ca2+ pathway implication during Xenopus laevis vitellogenesis.
Xepac protein and IP3/Ca2+ pathway implication during Xenopus laevis vitellogenesis.
Zygote (Cambridge, England) (2013-07-31)
María de los Angeles Serrano, Melchor Emilio Luque, Sara Serafina Sánchez
ABSTRACT
The objective of this study was to elucidate the signalling pathways initiated by cAMP once inside the Xenopus laevis oocyte, where it triggers and maintains vitellogenin endocytic uptake. Our results showed the presence of Xepac transcripts at all stages of oogenesis and we demonstrated that a cAMP analogue that exclusively activates Xepac, 8-CPT, was able to rescue the endocytic activity in oocytes with uncoupled gap junctions. Inhibition experiments for the IP3/Ca2+ signalling pathway showed either a complete inhibition or a significant reduction of the vitellogenic process. These results were confirmed with the rescue capability of the A-23187 ionophore in those oocyte batches in which the IP3/Ca2+ pathway was inhibited. Taking our findings into account, we propose that the cAMP molecule binds Xepac protein enabling it to activate the IP3/Ca2+ pathway, which is necessary to start and maintain X. laevis vitellogenin uptake.
Product Description
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
DL-Norleucine
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
(+)-Biotin N-hydroxysuccinimide ester, ≥98% (HPLC), powder
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Aminocaproic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
USP
Aminocaproic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Supelco
Aminocaproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)