Merck
COPD: early diagnosis and treatment to slow disease progression.
International journal of clinical practice (2014-11-05)
T Welte, C Vogelmeier, A Papi
ABSTRACT
Chronic obstructive pulmonary disease (COPD) is usually a progressive condition. Undiagnosed early-stage disease, particularly in symptomatic patients, is likely to become more severe with time. Hence, prevention or reduction in disease progression is highly relevant. We evaluated the published data and discussed the potential impact of early intervention on the course of COPD. We performed PubMed searches of studies in early or mild COPD, focusing on those relating to lung function decline. Smoking cessation reduced lung function decline at all stages of COPD, and the earlier the intervention, the greater the impact on lung function. Accumulating data from placebo-controlled trials suggested that long-acting bronchodilators can slow the decline in lung function, as well as reduce exacerbation and mortality rates and improve health-related quality of life (HRQoL) in patients with mild-to-moderate COPD. Inhaled corticosteroids (ICS) do not impact lung function in early COPD, and further research is needed on the role of long-acting β2-agonist-ICS combination therapy in these patients. Initiating treatment early in the course of COPD is likely to slow disease progression and improve HRQoL. Current data support maintenance treatment with a long-acting bronchodilator in this patient group. However, many questions remain unanswered regarding the optimal treatment of mild COPD, and further research is required to develop evidence-based recommendations in this field.
Product Description
USP
Fluticasone propionate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fluticasone propionate, ≥98% (HPLC), powder
Fluticasone propionate, European Pharmacopoeia (EP) Reference Standard