Merck
  • Evolutionary Action Score of TP53 Coding Variants Is Predictive of Platinum Response in Head and Neck Cancer Patients.
Evolutionary Action Score of TP53 Coding Variants Is Predictive of Platinum Response in Head and Neck Cancer Patients.
Cancer research (2015-02-19)
Abdullah A Osman, David M Neskey, Panagiotis Katsonis, Ameeta A Patel, Alexandra M Ward, Teng-Kuei Hsu, Stephanie C Hicks, Thomas O McDonald, Thomas J Ow, Marcus Ortega Alves, Curtis R Pickering, Heath D Skinner, Mei Zhao, Eric M Sturgis, Merrill S Kies, Adel El-Naggar, Federica Perrone, Lisa Licitra, Paolo Bossi, Marek Kimmel, Mitchell J Frederick, Olivier Lichtarge, Jeffrey N Myers
ABSTRACT
TP53 is the most frequently altered gene in head and neck squamous cell carcinoma (HNSCC), with mutations occurring in over two thirds of cases; however, the predictive response of these mutations to cisplatin-based therapy remains elusive. In the current study, we evaluate the ability of the Evolutionary Action score of TP53-coding variants (EAp53) to predict the impact of TP53 mutations on response to chemotherapy. The EAp53 approach clearly identifies a subset of high-risk TP53 mutations associated with decreased sensitivity to cisplatin both in vitro and in vivo in preclinical models of HNSCC. Furthermore, EAp53 can predict response to treatment and, more importantly, a survival benefit for a subset of head and neck cancer patients treated with platinum-based therapy. Prospective evaluation of this novel scoring system should enable more precise treatment selection for patients with HNSCC.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Serine, European Pharmacopoeia (EP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-p21WAF1 (Ab-1) Mouse mAb (EA10), liquid, clone EA10, Calbiochem®
Sigma-Aldrich
DL-Serine, ≥98% (TLC)