Merck
  • PD-1 is not required for natural or peripherally induced regulatory T cells: Severe autoimmunity despite normal production of regulatory T cells.
PD-1 is not required for natural or peripherally induced regulatory T cells: Severe autoimmunity despite normal production of regulatory T cells.
European journal of immunology (2014-09-23)
Kristofor K Ellestad, Govindarajan Thangavelu, Catherine L Ewen, Louis Boon, Colin C Anderson
ABSTRACT
The expression of the coinhibitor PD-1 on T cells is important for the establishment of immune homeostasis. We previously found that PD-1 is particularly critical for the control of self-tolerance during lymphopenia-induced proliferation of recent thymic emigrants (RTEs). Previous studies suggested that PD-1 modulates the generation of Treg cells, particularly peripherally induced Treg (pTreg) cells, and controls Th17 cells. However, these conclusions were derived indirectly from studies on the ligand PD-L1, and not PD-1 itself. Herein we directly tested whether T-cell PD-1 expression was needed for Treg cell generation and examined if a paucity of Treg cells or enhanced Th17 cells could explain the severe lymphopenia-potentiated autoimmunity caused by PD-1 KO RTEs. Employing the murine FoxP3(EGFP) reporter system to simultaneously monitor conversion of WT and PD-1 KO T cells to pTreg cells in the same animal, we found that PD-1 deficiency did not inhibit pTreg cell generation or lead to Th17-cell-mediated autoimmunity. Surprisingly, pTreg cell numbers were increased in PD-1 KO versus WT cell populations. Furthermore, we noted an increased conversion to pTreg cells by RTEs. Our data suggest that the primary role for PD-1 is to restrain T-cell activation/proliferation to self-Ags rather than promote generation of Treg cells.
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
Ampicillin, analytical standard