Merck
  • Cycloalkanones. 9. Comparison of analogues which inhibit cholesterol and fatty acid synthesis.
Cycloalkanones. 9. Comparison of analogues which inhibit cholesterol and fatty acid synthesis.
Journal of medicinal chemistry (1976-10-01)
I H Hall, G L Carlson
PMID11347
ABSTRACT
A number of 2,8-dibenzylcyclooctanone analogues inhibited the HMG-CoA reductases activity of Holtzman male rat liver, whereas only 2-octanone, 2-hexadecanone, 2,8-dibenzylcyclooctanone derivatives, and 2-bis(4-chlorophenyl)-3,5-dimethyltetrahydro-4-pyrone inhibited fatty acid synthetase activity. 2-Octanone significantly lowered serum cholesterol, triglycerides, and glycerol levels in Holtzman male rats and serlm cholesterol in male CF1 mice. Serum lipase activity was significantly elevated in rats administered 20 mg/kg/day of 2-octanone for 16 days. The activity of liver HMG-CoA reductase was inhibited in mice administered 10 mg/kg/day of 2-octanone for 10 days and in mouse and rat liver in vitro by 10 mg of 2-octanone. In mice, fecal excretion of [14C]cholesterol and tripalmitin was accelerated whereas palmitic acid and cholesteryl oleate were not affected by 10 mg/kg/day of 2-octanone. The LD50 in male mice for 2-octanone was 1.6g/kg.
Sigma-Aldrich
2-Hexadecanone, 98%
Pricing and availability is not currently available.