Merck
  • Resting potential of rat cerebellar granule cells during early maturation in vitro.
Resting potential of rat cerebellar granule cells during early maturation in vitro.
Journal of neurobiology (1997-01-01)
U Becherer, J L Rodeau, A Feltz
ABSTRACT
The survival of rat cerebellar granule cells maintained in vitro is enhanced by a KCl-enriched medium. This effect is classically interpreted as resulting from a higher cytosolic calcium concentration. This implies the presence of voltage-dependent Ca2+ channels and a membrane potential that can respond to changes in external K+. Since previous studies cast a doubt on these two conditions, we reinvestigated the resting membrane potential and Ca2+ influxes in rat cerebellar granule neurones during the first week in vitro using a fluorescence imaging approach. Membrane potential was assessed with the fluorescent dye bis-oxonol, and intracellular free calcium with Fura-2. Resting potential was shown to progressively decrease from -40 mV at the first day in vitro to -60 mV at day 7. At all times in culture, as early as day 0, cells were depolarized when external KCl concentration was increased from 5 to 30 mM. This depolarization resulted in an increased cytosolic calcium concentration due to Ca2+ influx through L-type and N-type voltage-activated Ca2+ channels, functional at day 0. Gross estimations of the permeabilities of Na+ and Cl- were obtained at various times in culture by measuring the changes in resting potential brought about by a reduction of their external concentration. A progressive increase of the relative permeability to K+ ions seems to underlie the evolution of the resting potential with time.
Sigma-Aldrich
DISBAC2(3), Voltage Sensitive Probe, ≥98% (HPLC)