- New developments in the pharmacologic treatment of obesity.
New developments in the pharmacologic treatment of obesity.
Nursing for women's health (2013-02-13)
Heidi Collins Fantasia
PMID23399013
ABSTRACT
Obesity, defined as a body mass index (BMI, kg/m(2) ) >30, is a significant public health problem. It's estimated that 50 percent of the U.S. population will be classified as obese by the year 2030. Due to associated health complications and rising health care costs related to obesity, new treatment options are being explored. For people who need additional treatment beyond lifestyle modification, new pharmacologic options have been developed that may assist in reducing BMI. Health care providers and patients should consider each person's individual health history and consider both the potential risks and benefits of these therapies.
Product Description
Supelco
Phentermine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®