Merck
  • Novel skeleton transformation reaction of α-pyrone derivatives to spirobicyclo[3.1.0]hexane derivatives using dimethylsulfoxonium methylide.
Novel skeleton transformation reaction of α-pyrone derivatives to spirobicyclo[3.1.0]hexane derivatives using dimethylsulfoxonium methylide.
Organic letters (2012-11-21)
Takuya Miura, Navnath Dnyanoba Yadav, Hiroki Iwasaki, Minoru Ozeki, Naoto Kojima, Masayuki Yamashita
ABSTRACT
By applying a skeleton transformation reaction using dimethylsulfoxonium methylide, a novel reaction was identified by which 5,6,7,8-tetrahydrocoumarin with the electron-withdrawing group at C3 was led to the spirobicyclo[3.1.0]hexane-cyclohexane derivative. Moreover, by establishing the scope of this reaction, it was confirmed that it is possible to apply this reaction to not only ring-fused α-pyrone derivatives but also alkyl-chain-substituted α-pyrone derivatives in moderate to good yields.
Product Description
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane-1-13C, 99 atom % 13C
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)