Merck
  • Effects of lanthanum chloride on glutamate level, intracellular calcium concentration and caspases expression in the rat hippocampus.
Effects of lanthanum chloride on glutamate level, intracellular calcium concentration and caspases expression in the rat hippocampus.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2012-10-26)
Jinghua Yang, Qiufang Liu, Shengwen Wu, Qi Xi, Yuan Cai
ABSTRACT
Lanthanum chloride (LaCl(3)) can affect neurobehavioral development and impair cognitive abilities. The mechanism underlying LaCl(3)-induced neurotoxic effects is still unknown. The purpose of this research was to investigate the neuronal impairment induced by LaCl(3) and discuss the possible mechanism from the aspects of the alteration of glutamate level, intracellular calcium concentration ([Ca(2+)](i)), Bax, Bcl-2 and caspases expression in the hippocampus. Lactational rats were exposed to 0, 0.25, 0.50 and 1.0 % LaCl(3) in drinking water, respectively. Their offspring were exposed to LaCl(3) by parental lactation and then administrated with 0, 0.25, 0.50 and 1.0 % LaCl(3) in drinking water for 1 month. The results showed that 0.25, 0.50 and 1.0 % LaCl(3) exposure induced neuronal impairment in the hippocampus of young rat. Hippocampal glutamate level, [Ca(2+)](i) and ratio of Bax and Bcl-2 expression increased significantly after LaCl(3) exposure. Besides, LaCl(3) exposure increased GRP78, GRP94, GADD153 and p-JNK expression, promoted the activation of caspase-3, caspase-9 and caspase-12, induced PARP cleavage and caused excessive apoptosis. These results indicate that LaCl(3) increases glutamate level, [Ca(2+)](i) and ratio of Bax and Bcl-2 expression, which cause excessive apoptosis by the mitochondrial and endoplasmic reticulum stress-induced pathway, and thus neuronal damages in the hippocampus.
Sigma-Aldrich
Lanthanum(III) chloride heptahydrate, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Lanthanum(III) chloride heptahydrate, ACS reagent
Sigma-Aldrich
Lanthanum(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis