Merck
  • Hair MDMA samples are consistent with reported ecstasy use: findings from a study investigating effects of ecstasy on mood and memory.
Hair MDMA samples are consistent with reported ecstasy use: findings from a study investigating effects of ecstasy on mood and memory.
Neuropsychobiology (2010-10-22)
A B Scholey, L Owen, J Gates, J Rodgers, T Buchanan, J Ling, T Heffernan, P Swan, C Stough, A C Parrott
ABSTRACT
Our group has conducted several Internet investigations into the biobehavioural effects of self-reported recreational use of MDMA (3,4-methylenedioxymethamphetamine or Ecstasy) and other psychosocial drugs. Here we report a new study examining the relationship between self-reported Ecstasy use and traces of MDMA found in hair samples. In a laboratory setting, 49 undergraduate volunteers performed an Internet-based assessment which included mood scales and the University of East London Drug Use Questionnaire, which asks for history and current drug use. They also provided a hair sample for determination of exposure to MDMA over the previous month. Self-report of Ecstasy use and presence in hair samples were consistent (p < 0.00001). Both subjective and objective measures predicted lower self-reported ratings of happiness and higher self-reported stress. Self-reported Ecstasy use, but not presence in hair, was also associated with decreased tension. Different psychoactive drugs can influence long-term mood and cognition in complex and dynamically interactive ways. Here we have shown a good correspondence between self-report and objective assessment of exposure to MDMA. These data suggest that the Internet has potentially high utility as a useful medium to complement traditional laboratory studies into the sequelae of recreational drug use.
Product Description
Supelco
(±)-MDEA solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(±)-3,4-Methylenedioxy-N-­ethyl­amphetamine hydrochloride, ≥98% (TLC)