Merck
  • Synthesis, structure and cytotoxicity of triphenylphosphinegold(I) sulfanylpropenoates.
Synthesis, structure and cytotoxicity of triphenylphosphinegold(I) sulfanylpropenoates.
Journal of inorganic biochemistry (2007-09-18)
Elena Barreiro, José S Casas, María D Couce, Agustín Sánchez, Angeles Sánchez-Gonzalez, José Sordo, José M Varela, Ezequiel M Vázquez López
ABSTRACT
The reaction of triphenylphosphinegold(I) chloride in ethanol in a 1:1 molar ratio with the 3-(aryl)-2-sulfanylpropenoic acids H(2)xspa [x: p=3-phenyl-, Clp=3-(2-chlorophenyl)-, -o-mp=3-(2-methoxyphenyl)-, -p-mp=3-(4-methoxyphenyl)-, -o-hp=3-(2-hydroxyphenyl)-, -p-hp=3-(4-hydroxyphenyl)-, diBr-o-hp=3-(3,5-dibromo-2-hydroxyphenyl)-, f=3-(2-furyl)-, t=3-(2-thienyl)-, -o-py=3-(2-pyridyl)-; spa=2-sulfanylpropenoato] gave compounds of the type [Au(PPh(3))(Hxspa)], which were isolated and characterized as solids by elemental analysis, IR spectroscopy and FAB mass spectrometry and in solution by (1)H, (13)C and (31)P NMR spectroscopy. The structures of the complexes [Au(PPh(3))(HClpspa)], [Au(PPh(3))(H-o-mpspa)] and [Au(PPh(3))(H-p-mpspa)].2/3C(3)H(6)O were determined by X-ray diffractometry. Hydrogen bonding was found along with Au-S and Au-P bonds in all cases and weak pi-pi stacking was found in the H-p-mpspa derivative. The in vitro antitumour activities against the HeLa-229, A2780 and A2780cis cell lines were determined for all complexes.
Sigma-Aldrich
Chloro(triphenylphosphine)gold(I), ≥99.9% trace metals basis