Merck
  • A Trojan horse transition state analogue generated by MgF3- formation in an enzyme active site.
A Trojan horse transition state analogue generated by MgF3- formation in an enzyme active site.
Proceedings of the National Academy of Sciences of the United States of America (2006-09-23)
Nicola J Baxter, Luis F Olguin, Marko Golicnik, Guoqiang Feng, Andrea M Hounslow, Wolfgang Bermel, G Michael Blackburn, Florian Hollfelder, Jonathan P Waltho, Nicholas H Williams
ABSTRACT
Identifying how enzymes stabilize high-energy species along the reaction pathway is central to explaining their enormous rate acceleration. beta-Phosphoglucomutase catalyses the isomerization of beta-glucose-1-phosphate to beta-glucose-6-phosphate and appeared to be unique in its ability to stabilize a high-energy pentacoordinate phosphorane intermediate sufficiently to be directly observable in the enzyme active site. Using (19)F-NMR and kinetic analysis, we report that the complex that forms is not the postulated high-energy reaction intermediate, but a deceptively similar transition state analogue in which MgF(3)(-) mimics the transferring PO(3)(-) moiety. Here we present a detailed characterization of the metal ion-fluoride complex bound to the enzyme active site in solution, which reveals the molecular mechanism for fluoride inhibition of beta-phosphoglucomutase. This NMR methodology has a general application in identifying specific interactions between fluoride complexes and proteins and resolving structural assignments that are indistinguishable by x-ray crystallography.
Sigma-Aldrich
Magnesium fluoride, technical grade
Sigma-Aldrich
Magnesium fluoride, random crystals, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium fluoride, pieces, 3-6 mm, 99.9% trace metals basis (excluding Na)