Merck
  • Adsorption of Bismark Brown dye on activated carbons prepared from rubberwood sawdust (Hevea brasiliensis) using different activation methods.
Adsorption of Bismark Brown dye on activated carbons prepared from rubberwood sawdust (Hevea brasiliensis) using different activation methods.
Journal of hazardous materials (2005-07-26)
B G Prakash Kumar, Lima Rose Miranda, M Velan
ABSTRACT
Hevea brasiliensis or rubberwood tree, as it is commonly known finds limited use once the latex has been tapped. The sawdust of this tree is chosen to ascertain it viability as a precursor for activation. The carbons thus obtained were characterized in terms of iodine, methylene blue number and surface area. The best carbon in each method was utilized to study the adsorption of Bismark Brown, a dye used in the leather industry. Adsorption equilibrium studies were carried out with the synthetic solutions of the dye, at room temperature (298 K). Equilibrium data are fitted with the Langmuir and the Freundlich isotherms models for the system. The effects of contact time, adsorbent dosage and initial dye concentrations on sorption capacity were carried out. Excellent adsorption capacities of 2000 and 1111 mg g(-1) were obtained for steam and chemical followed by steam-activated carbons, respectively. Pilot-plant experimental studies have been performed using packed-bed column with different feed concentrations, flow rates and bed heights, to evaluate sorption of Bismark Brown on steam-activated carbon. Bed depth service time (BDST) design model have been used to analyze the data.
Sigma-Aldrich
Bismarck Brown Y, certified by the BSC, Dye content 50 %