Merck
  • Dihydrooxonate is a substrate of dihydroorotate dehydrogenase (DHOD) providing evidence for involvement of cysteine and serine residues in base catalysis.
Dihydrooxonate is a substrate of dihydroorotate dehydrogenase (DHOD) providing evidence for involvement of cysteine and serine residues in base catalysis.
Archives of biochemistry and biophysics (2001-07-05)
O Björnberg, D B Jordan, B A Palfey, K F Jensen
ABSTRACT
The flavoprotein dihydroorotate dehydrogenase (DHOD) catalyzes the oxidation of dihydroorotate to orotate. Dihydrooxonate is an analogue of dihydroorotate in which the C5 carbon is substituted by a nitrogen atom. We have investigated dihydrooxonate as a substrate of three DHODs, each representing a distinct evolutionary class of the enzyme, namely the two family 1 enzymes from Lactococcus lactis, DHODA and DHODB, and the enzyme from Escherichia coli, which, like the human enzyme, belongs to family 2. Dihydrooxonate was accepted as a substrate although much less efficiently than dihydroorotate. The first half-reaction was rate limiting according to pre-steady-state and steady-state kinetics with different electron acceptors. Cysteine and serine have been implicated as active site base residues, which promote substrate oxidation in family 1 and family 2 DHODs, respectively. Mutants of DHODA (C130A) and E. coli DHOD (S175A) have extremely low activity in standard assays with dihydroorotate as substrate, but with dihydrooxonate the mutants display considerable and increasing activity above pH 8.0. Thus, the absence of the active site base residue in the enzymes seems to be compensated for by a lower pK(a) of the 5-position in the substrate. Oxonate, the oxidation product of dihydrooxonate, was a competitive inhibitor versus dihydroorotate, and DHODA was the most sensitive of the three enzymes. DHODA was reinvestigated with respect to product inhibition by orotate. The results suggest a classical one-site ping-pong mechanism with fumarate as electron acceptor, while the kinetics with ferricyanide is highly dependent on the detailed reaction conditions.
Sigma-Aldrich
D-Hydroorotic acid, 98%
Sigma-Aldrich
L-Dihydroorotic acid, ≥99%