Merck
  • Dominant role of the sst1 locus in pathogenesis of necrotizing lung granulomas during chronic tuberculosis infection and reactivation in genetically resistant hosts.
Dominant role of the sst1 locus in pathogenesis of necrotizing lung granulomas during chronic tuberculosis infection and reactivation in genetically resistant hosts.
The American journal of pathology (2009-05-16)
Alexander V Pichugin, Bo-Shiun Yan, Alex Sloutsky, Lester Kobzik, Igor Kramnik
ABSTRACT
Significant host heterogeneity in susceptibility to tuberculosis exists both between and within mammalian species. Using a mouse model of infection with virulent Mycobacterium tuberculosis (Mtb), we identified the genetic locus sst1 that controls the progression of pulmonary tuberculosis in immunocompetent hosts. In this study, we demonstrate that within the complex, multigenic architecture of tuberculosis susceptibility, sst1 functions to control necrosis within tuberculosis lesions in the lungs; this lung-specific sst1 effect is independent of both the route of infection and genetic background of the host. Moreover, sst1-dependent necrosis was observed at low bacterial loads in the lungs during reactivation of the disease after termination of anti-tuberculosis drug therapy. We demonstrate that in sst1-susceptible hosts, nonlinked host resistance loci control both lung inflammation and production of inflammatory mediators by Mtb-infected macrophages. Although interactions of the sst1-susceptible allele with genetic modifiers determine the type of the pulmonary disease progression, other resistance loci do not abolish lung necrosis, which is, therefore, the core sst1-dependent phenotype. Sst1-susceptible mice from tuberculosis-resistant and -susceptible genetic backgrounds reproduce a clinical spectrum of pulmonary tuberculosis and may be used to more accurately predict the efficacy of anti-tuberculosis interventions in genetically heterogeneous human populations.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Interleukin-3 from mouse, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Auramine O, Dye content ≥80 %, certified by the BSC
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Auramine O, Dye content 85 %, certified by the BSC
Sigma-Aldrich
Interleukin-3 human, IL-3, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Interleukin-3 human, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Sigma-Aldrich
Rhodamine B, for fluorescence
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Supelco
Rhodamine B, analytical standard