Merck
  • LC3-dependent extracellular vesicles promote M-MDSC accumulation and immunosuppression in colorectal cancer.
LC3-dependent extracellular vesicles promote M-MDSC accumulation and immunosuppression in colorectal cancer.
iScience (2024-05-06)
Ye Gu, Qiang Liu, Qiaoxian He, Qiangsheng Wu, Lingyun Li, Dongchao Xu, Liyun Zheng, Lu Xie, Sile Cheng, Hongzhang Shen, Yifeng Zhou, Jianfeng Yang, Hangbin Jin, Xiaofeng Zhang
ABSTRACT
For a long time, myeloid-derived suppressor cells (MDSCs) dilated in circulation system of colorectal cancer (CRC) patients have been puzzling clinicians. Various evidence shows that MDSCs constitute the bulk of immunosuppression in CRC, which is related to tumor growth, adhesion, invasion, metastasis, and immune escape. However, the mechanisms underlying these cells formation remain incompletely understood. In this study, we reported that CRC cell-derived LC3-dependent extracellular vesicles (LDEVs)-mediated M-MDSCs formation via TLR2-MYD88 pathway. Furthermore Hsp60 was the LDEVs surface ligand that triggered these MDSCs induction. In clinical studies, we reported that accumulation of circulating M-MDSCs as well as IL-10 and arginase1 secretion were reliant upon the levels of tumor cell-derived LDEVs in CRC patients. These findings indicated how local tumor cell-derived extracellular vesicles influence distal hematopoiesis and provided novel justification for therapeutic targeting of LDEVs in patients with CRC.
Sigma-Aldrich
Anti-Heat Shock Protein 70 (HSP70) antibody, Mouse monoclonal, clone BRM-22, purified from hybridoma cell culture
Sigma-Aldrich
Anti-HMGB1, affinity isolated antibody
Sigma-Aldrich
Anti-Podocalyxin (GP135) Antibody, clone 3F2:D8, clone 3F2:D8, from mouse
Sigma-Aldrich
Rabbit anti-HSP60 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Anti-HSP27 antibody produced in rabbit, affinity isolated antibody