Merck
  • C3aR-initiated signaling is a critical mechanism of podocyte injury in membranous nephropathy.
C3aR-initiated signaling is a critical mechanism of podocyte injury in membranous nephropathy.
JCI insight (2024-01-16)
Qi Zhang, Sofia Bin, Kelly Budge, Astgik Petrosyan, Valentina Villani, Paola Aguiari, Coralien Vink, Jack Wetzels, Hasmik Soloyan, Gaetano La Manna, Manuel Alfredo Podestà, Paolo Molinari, Sargis Sedrakyan, Kevin V Lemley, Roger E De Filippo, Laura Perin, Paolo Cravedi, Stefano Da Sacco
ABSTRACT
The deposition of antipodocyte autoantibodies in the glomerular subepithelial space induces primary membranous nephropathy (MN), the leading cause of nephrotic syndrome worldwide. Taking advantage of the glomerulus-on-a-chip system, we modeled human primary MN induced by anti-PLA2R antibodies. Here we show that exposure of primary human podocytes expressing PLA2R to MN serum results in IgG deposition and complement activation on their surface, leading to loss of the chip permselectivity to albumin. C3a receptor (C3aR) antagonists as well as C3AR gene silencing in podocytes reduced oxidative stress induced by MN serum and prevented albumin leakage. In contrast, inhibition of the formation of the membrane-attack-complex (MAC), previously thought to play a major role in MN pathogenesis, did not affect permselectivity to albumin. In addition, treatment with a C3aR antagonist effectively prevented proteinuria in a mouse model of MN, substantiating the chip findings. In conclusion, using a combination of pathophysiologically relevant in vitro and in vivo models, we established that C3a/C3aR signaling plays a critical role in complement-mediated MN pathogenesis, indicating an alternative therapeutic target for MN.
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (HPLC), buffered aqueous solution
Sigma-Aldrich
Anti-PLA2R Antibody, clone 5F5.1, clone 5F5.1, from mouse
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution