Merck
SiC Nanofibers as Long-Life Lithium-Ion Battery Anode Materials.
Frontiers in chemistry (2018-06-06)
Xuejiao Sun, Changzhen Shao, Feng Zhang, Yi Li, Qi-Hui Wu, Yonggang Yang
ABSTRACT
The development of high energy lithium-ion batteries (LIBs) has spurred the designing and production of novel anode materials to substitute currently commercial using graphitic materials. Herein, twisted SiC nanofibers toward LIBs anode materials, containing 92.5 wt% cubic β-SiC and 7.5 wt% amorphous C, were successfully synthesized from resin-silica composites. The electrochemical measurements showed that the SiC-based electrode delivered a stable reversible capacity of 254.5 mAh g-1 after 250 cycles at a current density of 0.1 A g-1. It is interesting that a high discharge capacity of 540.1 mAh g-1 was achieved after 500 cycles at an even higher current density of 0.3 A g-1, which is higher than the theoretical capacity of graphite. The results imply that SiC nanomaterials are potential anode candidate for LIBs with high stability due to their high structure stability as supported with the transmission electron microscopy images.
Sigma-Aldrich
Silicon carbide, nanopowder, ≥99% trace metals basis, 18 nm avg. part. size