Merck
  • Paracrine signal emanating from stressed cardiomyocytes aggravates inflammatory microenvironment in diabetic cardiomyopathy.
Paracrine signal emanating from stressed cardiomyocytes aggravates inflammatory microenvironment in diabetic cardiomyopathy.
iScience (2022-03-15)
Namrita Kaur, Andrea Ruiz-Velasco, Rida Raja, Gareth Howell, Jessica M Miller, Riham R E Abouleisa, Qinghui Ou, Kimberly Mace, Susanne S Hille, Norbert Frey, Pablo Binder, Craig P Smith, Helene Fachim, Handrean Soran, Eileithyia Swanton, Tamer M A Mohamed, Oliver J Müller, Xin Wang, Jonathan Chernoff, Elizabeth J Cartwright, Wei Liu
ABSTRACT
Myocardial inflammation contributes to cardiomyopathy in diabetic patients through incompletely defined underlying mechanisms. In both human and time-course experimental samples, diabetic hearts exhibited abnormal ER, with a maladaptive shift over time in rodents. Furthermore, as a cardiac ER dysfunction model, mice with cardiac-specific p21-activated kinase 2 (PAK2) deletion exhibited heightened myocardial inflammatory response in diabetes. Mechanistically, maladaptive ER stress-induced CCAAT/enhancer-binding protein homologous protein (CHOP) is a novel transcriptional regulator of cardiac high-mobility group box-1 (HMGB1). Cardiac stress-induced release of HMGB1 facilitates M1 macrophage polarization, aggravating myocardial inflammation. Therapeutically, sequestering the extracellular HMGB1 using glycyrrhizin conferred cardioprotection through its anti-inflammatory action. Our findings also indicated that an intact cardiac ER function and protective effects of the antidiabetic drug interdependently attenuated the cardiac inflammation-induced dysfunction. Collectively, we introduce an ER stress-mediated cardiomyocyte-macrophage link, altering the macrophage response, thereby providing insight into therapeutic prospects for diabetes-associated cardiac dysfunction.
Sigma-Aldrich
Vildagliptin, ≥98% (HPLC)
Sigma-Aldrich
Anti-Rac1 Antibody, from rabbit
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Cholesterol Quantitation Kit, sufficient for 100 colorimetric or fluorometric tests
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection