Merck
  • Fragment-Based Discovery of MRTX1719, a Synthetic Lethal Inhibitor of the PRMT5•MTA Complex for the Treatment of MTAP-Deleted Cancers.
Fragment-Based Discovery of MRTX1719, a Synthetic Lethal Inhibitor of the PRMT5•MTA Complex for the Treatment of MTAP-Deleted Cancers.
Journal of medicinal chemistry (2022-01-19)
Christopher R Smith, Ruth Aranda, Thomas P Bobinski, David M Briere, Aaron C Burns, James G Christensen, Jeffery Clarine, Lars D Engstrom, Robin J Gunn, Anthony Ivetac, Ronald Jean-Baptiste, John M Ketcham, Masakazu Kobayashi, Jon Kuehler, Svitlana Kulyk, J David Lawson, Krystal Moya, Peter Olson, Lisa Rahbaek, Nicole C Thomas, Xiaolun Wang, Laura M Waters, Matthew A Marx
ABSTRACT
The PRMT5•MTA complex has recently emerged as a new synthetically lethal drug target for the treatment of MTAP-deleted cancers. Here, we report the discovery of development candidate MRTX1719. MRTX1719 is a potent and selective binder to the PRMT5•MTA complex and selectively inhibits PRMT5 activity in MTAP-deleted cells compared to MTAP-wild-type cells. Daily oral administration of MRTX1719 to tumor xenograft-bearing mice demonstrated dose-dependent inhibition of PRMT5-dependent symmetric dimethylarginine protein modification in MTAP-deleted tumors that correlated with antitumor activity. A 4-(aminomethyl)phthalazin-1(2H)-one hit was identified through a fragment-based screen, followed by X-ray crystallography, to confirm binding to the PRMT5•MTA complex. Fragment growth supported by structural insights from X-ray crystallography coupled with optimization of pharmacokinetic properties aided the discovery of development candidate MRTX1719.
Sigma-Aldrich
Anti-dimethyl-Arginine Antibody, symmetric (SYM11), Upstate®, from rabbit