Merck
  • Phorbol-12-myristate 13-acetate acting through protein kinase Cepsilon induces translocator protein (18-kDa) TSPO gene expression.
Phorbol-12-myristate 13-acetate acting through protein kinase Cepsilon induces translocator protein (18-kDa) TSPO gene expression.
Biochemistry (2008-11-04)
Amani Batarseh, Christoforos Giatzakis, Vassilios Papadopoulos
ABSTRACT
Translocator protein (TSPO) is an 18-kDa cholesterol-binding protein that is expressed at high levels in steroid synthesizing and several cancer cells where it is involved in steroidogenesis and cell proliferation, respectively. The factors regulating Tspo expression are unknown. We analyzed Tspo transcriptional responses to the tumor promoter, phorbol-12-myristate 13-acetate (PMA), in cells with varying TSPO levels. PMA induced Tspo promoter activity and Tspo mRNA levels in TSPO-poor nonsteroidogenic cells (NIH-3T3 fibroblasts and COS-7 kidney) but not in TSPO-rich steroidogenic cells (MA-10 Leydig) with high basal Tspo transcriptional activity. The stimulatory effect of PMA was mediated by an 805-515-bp region upstream of the transcription start site. Electrophoretic mobility shift assay (EMSA) revealed that PMA induced binding of c-jun and GA-binding protein transcription factor (GABP-alpha) to their respective activator protein 1 (AP1) and v-ets erythroblastosis virus E26 oncogene homologue (Ets) sites in this region. Protein kinase C (PKC)-specific inhibitors blocked PMA induction of Tspo promoter activity with an inhibition profile suggestive of involvement of PKCepsilon. PKCepsilon expression correlated with TSPO content in the three cell lines. In NIH-3T3 cells, PKCepsilon overexpression induced Tspo promoter activity and mRNA levels and enhanced PMA-induced up regulation of c-jun and TSPO. In MA-10 cells, a PKCepsilon-specific translocation inhibitor peptide reduced basal Tspo promoter activity. PKCepsilon siRNA pool reduced PKCepsilon and TSPO levels in MA-10 cells indicating a role for PKCepsilon in regulating TSPO expression. Taken together, these data suggest that elevated TSPO expression in steroidogenic cells may be due to high constitutive expression of PKCepsilon that renders them unresponsive to further induction while PMA activation of PKCepsilon drives inducible TSPO expression in nonsteroidogenic cells, likely through AP1 and Ets.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
4α-Phorbol 12-myristate 13-acetate, solid, ≥95% (TLC)