Merck
  • Discovery and Characterization of BAY 1214784, an Orally Available Spiroindoline Derivative Acting as a Potent and Selective Antagonist of the Human Gonadotropin-Releasing Hormone Receptor as Proven in a First-In-Human Study in Postmenopausal Women.
Discovery and Characterization of BAY 1214784, an Orally Available Spiroindoline Derivative Acting as a Potent and Selective Antagonist of the Human Gonadotropin-Releasing Hormone Receptor as Proven in a First-In-Human Study in Postmenopausal Women.
Journal of medicinal chemistry (2020-09-23)
Olaf Panknin, Andrea Wagenfeld, Wilhelm Bone, Eckhard Bender, Katrin Nowak-Reppel, Amaury E Fernández-Montalván, Reinhard Nubbemeyer, Stefan Bäurle, Sven Ring, Norbert Schmees, Olaf Prien, Martina Schäfer, Christian Friedrich, Thomas M Zollner, Andreas Steinmeyer, Thomas Mueller, Gernot Langer
ABSTRACT
The growth of uterine fibroids is sex hormone-dependent and commonly associated with highly incapacitating symptoms. Most treatment options consist of the control of these hormonal effects, ultimately blocking proliferative estrogen signaling (i.e., oral contraceptives/antagonization of human gonadotropin-releasing hormone receptor [hGnRH-R] activity). Full hGnRH-R blockade, however, results in menopausal symptoms and affects bone mineralization, thus limiting treatment duration or demanding estrogen add-back approaches. To overcome such issues, we aimed to identify novel, small-molecule hGnRH-R antagonists. This led to the discovery of compound BAY 1214784, an orally available, potent, and selective hGnRH-R antagonist. Altering the geminal dimethylindoline core of the initial hit compound to a spiroindoline system significantly improved GnRH-R antagonist potencies across several species, mandatory for a successful compound optimization in vivo. In a first-in-human study in postmenopausal women, once daily treatment with BAY 1214784 effectively lowered plasma luteinizing hormone levels by up to 49%, at the same time being associated with low pharmacokinetic variability and good tolerability.
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
BAY-786, ≥98% (HPLC)
Sigma-Aldrich
BAY-784, ≥98% (HPLC)
Sigma-Aldrich
Luteinizing hormone releasing hormone human acetate salt, ≥98% (HPLC), powder