Merck
  • Serum immunoglobulin G antibody to Porphyromonas gingivalis in rapidly progressive periodontitis: titer, avidity, and subclass distribution.
Serum immunoglobulin G antibody to Porphyromonas gingivalis in rapidly progressive periodontitis: titer, avidity, and subclass distribution.
Infection and immunity (1992-06-01)
C Whitney, J Ant, B Moncla, B Johnson, R C Page, D Engel
ABSTRACT
Porphyromonas gingivalis is a suspected pathogen in rapidly progressive periodontitis (RPP). We have determined the anti-P. gingivalis serum immunoglobulin G (IgG) isotype response and avidity and the subclass titer distributions for 30 RPP patients and 30 age-, sex-, and race-matched healthy subjects by using enzyme-linked immunosorbent assay technology. Patients and control subjects were classified as seropositive if their total IgG response to P. gingivalis was twofold or more than the median response in healthy subjects. The predominant antibody responses for both patients and healthy subjects were IgG2 and IgG3, with a subclass order of IgG2 greater than IgG3 greater than IgG1 greater than IgG4. The avidity of the IgG response was highest for the seropositive healthy subjects and was no different between seronegative and seropositive RPP patients. The subclass antibody responses did not depend on gender, and there were no correlations between titer, avidity, or subclass with disease severity in the RPP patients as measured by pocket depth or bone loss on dental X rays. The seronegative RPP patients exhibited antibody responses that were greater than the responses of seronegative healthy subjects for all four subclasses, while the seropositive RPP patients had higher IgG1 and IgG4 levels than seropositive healthy subjects. These findings are consistent with the hypothesis that both carbohydrate and protein antigens are important in the IgG response to P. gingivalis. The relative predominance of IgG2, a subclass which lacks strong complement fixation and opsonic properties, and the low avidity of patient anti-P. gingivalis IgG antibodies suggest that humoral responsiveness to infection with P. gingivalis may be ineffective in clearing this organism.
Sigma-Aldrich
Anti-Human IgG1−Biotin antibody, Mouse monoclonal, clone 8c/6-39, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG3−Biotin antibody, Mouse monoclonal, clone HP-6050, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG2−Biotin antibody, Mouse monoclonal, clone HP-6014, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG4−Biotin antibody, Mouse monoclonal, clone HP-6025, purified from hybridoma cell culture