Merck
  • Sorbus umbellata (Desf.) Fritsch var. umbellata Leaves: Optimization of Extraction Conditions and Investigation Antimicrobial, Cytotoxic, and β-Glucuronidase Inhibitory Potential.
Sorbus umbellata (Desf.) Fritsch var. umbellata Leaves: Optimization of Extraction Conditions and Investigation Antimicrobial, Cytotoxic, and β-Glucuronidase Inhibitory Potential.
Plant foods for human nutrition (Dordrecht, Netherlands) (2019-06-20)
Dilek Demirbuker Kavak, Bilge Akdeniz
ABSTRACT
This study aimed to optimize the extraction conditions for Sorbus umbellata (Desf.) Fritsch var. umbellata leaves to maximize the phenolic content and their antioxidant activity and to investigate β-glucuronidase (GUS) enzyme inhibitory, antimicrobial and cytotoxic potentials of the extracts obtained under optimum conditions. The optimum extraction conditions were found to be 78.2 and 79.7% solvent, 73.1 and 71.5 °C, and 89.9 and 88.8 min to maximize phenolic content and antioxidant activity, respectively. Low values of coefficient of variations indicate the high reliability and reproducibility of the conducted extraction experiments. Bioactivity results showed that extracts had cytotoxic effect on the MCF-7 and A549 cells where the highest cell proliferation inhibition was observed for the A549 cell line (71.8% at 150 μg/mL). Staphylococcus aureus showed highest zone of inhibition (19.3 mm) in all bacteria followed by Escherichia coli. Additionally, extracts displayed potential GUS inhibitory activity. In conclusion, Sorbus umbellata leaf extract can be obtained by optimized cost-saving extraction and has a potential bioactivity to be utilized as a food ingredient for high value-added products and/or nutraceuticals development where it can combat oxidative stress and GUS mediated reactive metabolite formation.