- Effects of the Chiral Fungicides Metalaxyl and Metalaxyl-M on the Earthworm Eisenia fetida as Determined by ¹H-NMR-Based Untargeted Metabolomics.
Effects of the Chiral Fungicides Metalaxyl and Metalaxyl-M on the Earthworm Eisenia fetida as Determined by ¹H-NMR-Based Untargeted Metabolomics.
Although metalaxyl and metalaxyl-M are widely used fungicides, very little is known about their subacute and enantiospecific effects on the earthworm metabolome. In this study, Eisenia fetida were exposed to metalaxyl and metalaxyl-M at three concentrations (0.5, 5 and 50 mg/kg) for seven days. ¹H nuclear magnetic resonance (¹H-NMR)-based untargeted metabolomics showed that metalaxyl and metalaxyl-M exposure disturbed earthworms' metabolism at all three concentrations. Endogenous metabolites, such as succinate, arginine, aspartate, urea, asparagine, alanine, trimethylamine, taurine, cysteine, serine, threonine, histidine, lysine, glucose, choline, carnitine, citric acid, alpha-ketoisovaleric acid, fumaric acid and so on, were significantly changed. These results indicate that metalaxyl and metalaxyl-M produce different, enantiospecific disturbances in the earthworm metabolism, particularly in the tricarboxylic acid (TCA) and urea cycles. The application of untargeted metabolomics thus provides more information for evaluating the toxic risks of metalaxyl and metalaxyl-M.