Merck
  • Human colonic crypts in culture: segregation of immunochemical markers in normal versus adenoma-derived.
Human colonic crypts in culture: segregation of immunochemical markers in normal versus adenoma-derived.
Laboratory investigation; a journal of technical methods and pathology (2013-12-25)
Michael K Dame, Yan Jiang, Henry D Appelman, Kelly D Copley, Shannon D McClintock, Muhammad Nadeem Aslam, Durga Attili, B Joseph Elmunzer, Dean E Brenner, James Varani, D Kim Turgeon
ABSTRACT
In order to advance a culture model of human colonic neoplasia, we developed methods for the isolation and in vitro maintenance of intact colonic crypts from normal human colon tissue and adenomas. Crypts were maintained in three-dimensional Matrigel culture with a simple, serum-free, low Ca(2+) (0.15 mM) medium. Intact colonic crypts from normal human mucosa were viably maintained for 3-5 days with preservation of the in situ crypt-like architecture, presenting a distinct base and apex. Abnormal structures from adenoma tissue could be maintained through multiple passages (up to months), with expanding buds/tubules. Immunohistochemical markers for intestinal stem cells (Lgr5), growth (Ki67), differentiation (E-cadherin, cytokeratin 20 (CK20) and mucin 2 (MUC2)) and epithelial turnover (Bax, cleaved Caspase-3), paralleled the changes in function. The epithelial cells in normal crypts followed the physiological sequence of progression from proliferation to differentiation to dissolution in a spatially and temporally appropriate manner. Lgr5 expression was seen in a few basal cells of freshly isolated crypts, but was not detected after 1-3 days in culture. After 24 h in culture, crypts from normal colonic tissue continued to show strong Ki67 and MUC2 expression at the crypt base, with a gradual decrease over time such that by days 3-4 Ki67 was not expressed. The differentiation marker CK20 increased over the same period, eventually becoming intense throughout the whole crypt. In adenoma-derived structures, expression of markers for all stages of progression persisted for the entire time in culture. Lgr5 showed expression in a few select cells after months in culture. Ki67 and MUC2 were largely associated with the proliferative budding regions while CK20 was localized to the parent structure. This ex vivo culture model of normal and adenomatous crypts provides a readily accessible tool to help understand the growth and differentiation process in human colonic epithelium.
Sigma-Aldrich
Anti-E-Cadherin Antibody, clone 67A4, Azide Free, clone 67A4, Chemicon®, from mouse
Sigma-Aldrich
Goat Serum Donor Herd, USA origin, sterile-filtered, suitable for cell culture