Merck
  • Methyl group transfer upon gas phase decomposition of protonated methyl benzoate and similar compounds.
Methyl group transfer upon gas phase decomposition of protonated methyl benzoate and similar compounds.
Journal of mass spectrometry : JMS (2018-02-15)
Rafał Frański, Błażej Gierczyk, Maciej Zalas, Wojciech Jankowski, Marcin Hoffmann
ABSTRACT
Gas phase decompositions of protonated methyl benzoate and its conjugates have been studied by using electrospray ionization-collision induced dissociation-tandem mass spectrometry. Loss of CO2 molecule, thus transfer of methyl group, has been observed. In order to better understand this process, the theoretical calculations have been performed. For methyl benzoate conjugates, it has been found that position of substituent affects the loss of CO2 molecule, not the electron donor/withdrawing properties of the substituent. Therefore, electrospray ionization-mass spectrometry in positive ion mode may be useful for differentiation of isomers of methyl benzoate conjugates.
Sigma-Aldrich
Phenyl benzoate, 99%
Sigma-Aldrich
Methyl 4-fluorobenzoate, 98%
Sigma-Aldrich
Methyl 2-naphthoate, 97%