
Triphenylphosphine, polymer-bound

Synonym(s):
Copolymer of styrene and divinylbenzene, diphenylphosphinated, Diphenylphosphino-polystyrene, Polystyrene crosslinked with divinylbenzene, diphenylphosphinated
Linear Formula:
-C6H4P(C6H5)2
CAS Number:
Molecular Weight:
262.29
Products
Product Category
Brand
Formula Weight
Physical Form
Functional Group
Chain Type
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing