![Topramezone PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/289/476/aeaa851a-6b49-494f-b5bb-aa8abdd6cf61/640/aeaa851a-6b49-494f-b5bb-aa8abdd6cf61.png)
Topramezone
![Topramezone PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/289/476/aeaa851a-6b49-494f-b5bb-aa8abdd6cf61/640/aeaa851a-6b49-494f-b5bb-aa8abdd6cf61.png)
Synonym(s):
[3-(4,5-Dihydro-3-isoxazolyl)-2-methyl-4-(methylsulfonyl)phenyl](5-hydroxy-1-methyl-1H-pyrazol-4-yl)methanone
Empirical Formula (Hill Notation):
C16H17N3O5S
CAS Number:
Molecular Weight:
363.39
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing