![SRT® SEC-150 Gel Filtration Guard Column, 5 μm L × I.D. 5 cm × 7.8 mm, 5 μm particle size, stainless steel](/deepweb/assets/sigmaaldrich/product/images/328/678/90e144af-98c6-46a5-84d8-ec244ae90747/640/90e144af-98c6-46a5-84d8-ec244ae90747.jpg)
SRT® SEC-150 (5 µm) HPLC Columns
![SRT® SEC-150 Gel Filtration Guard Column, 5 μm L × I.D. 5 cm × 7.8 mm, 5 μm particle size, stainless steel](/deepweb/assets/sigmaaldrich/product/images/328/678/90e144af-98c6-46a5-84d8-ec244ae90747/640/90e144af-98c6-46a5-84d8-ec244ae90747.jpg)
Products
Product Category
Manufacturer
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing