Skip to Content
Merck
rac-Glycerol 1-myristate ≥99%

rac-Glycerol 1-myristate

rac-Glycerol 1-myristate ≥99%
Synonym(s):
1-Monotetradecanoyl-rac-glycerol, rac-1-Myristoylglycerol, DL-α-Myristin, Monomyristin
Linear Formula:
H3C(CH2)12COOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
302.45

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing