
Procymidone

Empirical Formula (Hill Notation):
C13H11Cl2NO2
Molecular Weight:
284.14
EC Number:
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing