
Pregabalin impurity B

Synonym(s):
(3R)-3-(Aminomethyl)-5-methylhexanoic acid
Empirical Formula (Hill Notation):
C8H17NO2
CAS Number:
Molecular Weight:
159.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing