Skip to Content
DJ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette tips in 384-tip format, robotic
Products
Product Category
pipette tips (7)
Manufacturer
Color
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
AXYVT38470ULR
Pipette tips in 384-tip format, robotic,
size 70 μL, pkg of 5x3840tips/cs, racked
Expand
View Pricing
AXYVT38470ULRS
Pipette tips in 384-tip format, robotic,
size 70 μL, pkg of 5x3840tips/cs, racked, sterile
Expand
View Pricing
AXYVT38410ULRS
Pipette tips in 384-tip format, robotic,
size 10 μL, pkg of 5x3840tips/cs, racked, sterile
Expand
View Pricing
AXYVT38431ULR
Pipette tips in 384-tip format, robotic,
size 30 μL, pkg of 5x3840tips/cs, racked
Expand
View Pricing
AXYVT3845ULR
Pipette tips in 384-tip format, robotic,
size 5 μL, pkg of 5x3840tips/cs, racked
Expand
View Pricing
AXYVT38410ULR
Pipette tips in 384-tip format, robotic,
size 10 μL, pkg of 5x3840tips/cs, racked
Expand
View Pricing
AXYVT38431ULRS
Pipette tips in 384-tip format, robotic,
size 30 μL, pkg of 5x3840tips/cs, racked, sterile
Expand
View Pricing