
Pantoprazole sodium sesquihydrate

Empirical Formula (Hill Notation):
C16H14F2N3NaO4S · 1.5H2O
CAS Number:
Molecular Weight:
432.37
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing