
Oseltamivir phosphate (impurity B free)

Synonym(s):
Oseltamivir phosphate, (3R,4R,5S)-4-Acetamido-5-amino-3-(1-ethylpropoxy)-1-cyclohexene-1-carboxylic acid ethyl ester phosphate salt
Empirical Formula (Hill Notation):
C16H28N2O4 ·H3PO4
CAS Number:
Molecular Weight:
410.40
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing