
Nadolol

Synonym(s):
Nadolol
Empirical Formula (Hill Notation):
C17H27NO4
CAS Number:
Molecular Weight:
309.40
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing