
Metoclopramide Related Compound B

Synonym(s):
Methyl 4-acetamido-5-chloro-2-methoxybenzoate
Linear Formula:
CH3CONHC6H2(Cl)(OCH3)CO2CH3
CAS Number:
Molecular Weight:
257.67
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2