
Menaquinone-7

Synonym(s):
Vitamin K2-(MK-7), (all-E)–2-(3,7,11,15,19,23,27-Heptamethyl-2,6,10,14,18,22,26-octacosaheptaenyl)-3-methyl-1,4-naphthalenedione
Empirical Formula (Hill Notation):
C46H64O2
CAS Number:
Molecular Weight:
649.00
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2