Medroxyprogesterone acetate for peak identification
Synonym(s):
Medroxyprogesterone 17-acetate, 17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate
Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing