
Lutein (Zeaxanthin free)

Synonym(s):
Xanthophyll, α-Carotene-3,3′-diol, β, ε-carotene, β, ε-carotene-3,3′-diol, Lutein
Empirical Formula (Hill Notation):
C40H56O2
CAS Number:
Molecular Weight:
568.87
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing