![Hydrochloric acid 10% EMPROVE® EXPERT Ph Eur,JP,NF](/deepweb/assets/sigmaaldrich/product/structures/546/395/10e67a13-e4c4-4e30-a1da-32e605ea8bfd/640/10e67a13-e4c4-4e30-a1da-32e605ea8bfd.png)
Hydrochloric acid 10%
![Hydrochloric acid 10% EMPROVE® EXPERT Ph Eur,JP,NF](/deepweb/assets/sigmaaldrich/product/structures/546/395/10e67a13-e4c4-4e30-a1da-32e605ea8bfd/640/10e67a13-e4c4-4e30-a1da-32e605ea8bfd.png)
Synonym(s):
Hydrochloric acid solution, Hydrogen chloride solution
Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Products
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing