
Gelatin methacryloyl

Synonym(s):
GelMa, Gelatin Methacrylate, Gelatin methacrylamide
Linear Formula:
(C40H59N11O13)n
Products
Brand
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-5 of 5
Compare
Product Number
Description
Pricing