
Fmoc-N-Me-Tyr(tBu)-OH

Synonym(s):
fmoc-N-me-tyr(tbu)-oh, Fmoc-N-Me-Tyr(tBu)-OH
Empirical Formula (Hill Notation):
C29H31NO5
CAS Number:
Molecular Weight:
473.56
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing