
Fmoc-Cys(tBu)-OH

Synonym(s):
Nα-Fmoc-S-tert-butyl-L-cysteine, Fmoc-Cys(tBu)-OH
Empirical Formula (Hill Notation):
C22H25NO4S
CAS Number:
Molecular Weight:
399.50
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing