
Fluoxetine Related Compound C

Synonym(s):
N-Methyl-N-[3-phenyl-3-(4-trifluoromethyl-phenoxy)-propyl]-succinamic acid
Empirical Formula (Hill Notation):
C21H22F3NO4
CAS Number:
Molecular Weight:
409.40
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2