
Flazasulfuron

Synonym(s):
N-(4,6-Dimethoxy-2-pyrimidinylaminocarbonyl)-3-(trifluoromethyl)-2-pyridinesulfonamide
Empirical Formula (Hill Notation):
C13H12F3N5O5S
CAS Number:
Molecular Weight:
407.33
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing