
Diclofop-methyl

Empirical Formula (Hill Notation):
C16H14Cl2O4
CAS Number:
Molecular Weight:
341.19
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing