
Deuteroporphyrin IX dimethyl ester from bovine blood

Synonym(s):
Dimethyl 3,7,12,17-tetramethyl-21H,23H-porphine-2,18-dipropionate, NSC 17451
Empirical Formula (Hill Notation):
C32H34N4O4
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing